CLS707710N
Pyrex® disposable serological pipettes
volume 10.0 mL
Synonym(s):
Corning® 7077
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
Recommended Products
material
borosilicate glass
sterility
sterile
feature
DNase free: no
RNase free: no
graduations
subdivision
packaging
pack of 100 ea
case of 600 ea
manufacturer/tradename
Corning 7077-10N
measuring range
accuracy: 0.1 mL
capacity
10.0 mL
volume
10.0 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Pyrex® disposable borosilcate glass serological pipettes are calibrated "to deliver" with blow-out.
- Features long, slender, tapered tips for faster and easier pipetting.
- Includes negative graduations for additional working volume.
- Fire-polished tips for burr-free, even flow rates.
These disposable serological pipettes are calibrated "to deliver" with blow -out. Individually wrapped. Sterile. Plugged
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service