CLS6795200
Corning® hotplate, 4 in. x 5 in.
AC/DC input 120 V AC, US 3-pin plug, plate L × W 4 in. × 5 in.
Synonym(s):
stirrers
About This Item
Recommended Products
material
ceramic plate
sterility
non-sterile
manufacturer/tradename
Corning 6795-200
availability
not available in EU
parameter
25-550 °C temp. range
plate L × W
4 in. × 5 in.
color
none
Looking for similar products? Visit Product Comparison Guide
Application
Quality
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service