Key Documents
CLS6383250
Pyrex® cylindrical separatory funnel, PTFE stopcock
Synonym(s):
Corning® 6383
Select a Size
4 EA
$578.00
1 EA
$699.00
$578.00
Estimated to ship onMarch 25, 2025
Select a Size
Change View
4 EA
$578.00
1 EA
$699.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
$578.00
Estimated to ship onMarch 25, 2025
Joint
joint stopper22
manufacturer/tradename
Corning 6383-250
capacity
250 mL
stopcock size
2
General description
Graduated, with ST Pyrex stopper and microfinish PTFE stopcock.
CLS6383A100 - does not include a stopper
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)