Key Documents
CLS606090
Pyrex® Buchner funnel, with perforated plate
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
manufacturer/tradename
Corning 6060-90
diam.
90.0 mm
color
clear
General description
With perforated plate. Suitable for vacuum filtration, but translucent plate should not be subjected to pressure differential in excess of 1atm.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)