Skip to Content
Merck
All Photos(1)

Key Documents

CLS563550

Pyrex® wide mouth micro volumetric flask with Pyrex® ST stopper

capacity 50 mL

Synonym(s):

Corning® 5635

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
$292.00

$292.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
6 EA
$292.00

About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

$292.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

borosilicate glass
wide-mouth

stopper size

13

sterility

non-sterile

feature

class A
closure type ST stopper

packaging

case of 6

manufacturer/tradename

Corning 5635-50

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

The heavy duty, wide mouth facilitates filling and mixing.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service