CLS43205L
Pyrex® round-bottom boiling flask, short neck
capacity 5,000 mL
Synonym(s):
bottom flask, flask glass, round flask
Select a Size
$986.00
Select a Size
About This Item
$986.00
Recommended Products
material
borosilicate glass
round bottom flask
short
Joint
joint ST/NS45/50
sterility
non-sterile
feature
closure type Standard taper joint
packaging
case of 4
manufacturer/tradename
Corning 4320-5L
availability
not available in EU
capacity
5,000 mL
neck size
(1)
Related Categories
General description
- PYREX® round-bottom boiling flasks are routinely used in various laboratory applications.
- Manufactured from low expansion borosilicate glass that provides better clarity, strength, heat, and chemical resistance even in the most demanding laboratory conditions.
- Precise wall thickness ensures optimal thermal transfer and provides mechanical strength for greater shock resistance.
- Chemically durable, these flasks are ideal for contamination-free results.
- The normal working temperature for PYREX® boling flasks is 230 °C.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)