Key Documents
CLS361125
Pyrex® moisture test distilling receiver, 24/40 joints
capacity 25 mL
Synonym(s):
Corning® 3611
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
manufacturer/tradename
Corning 3611-25
H
317 mm
capacity
25 mL
Related Categories
General description
Graduated, from 0 to 1 mL, in 0.1 mL intervals, with a maximum scale error of 0.05 mL. Graduation intervals from 1.0 to 10.0 mL are 0.2 mL, with a maximum scale error of 0.1 mL.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)