Skip to Content
Merck
All Photos(1)

Key Documents

CLS2962100

Pyrex® hydrometer jar

capacity 100 mL

Synonym(s):

Corning® 2962

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113024
NACRES:
NB.43
Pricing and availability is not currently available.

manufacturer/tradename

Corning 2962-100

O.D. × H

29 mm × 254 mm

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

Breakage is minimized by the heavy wall construction. The large hexagonal base, sealed to the cylinder body, increases stability. Meets ASTM specifications D-287 and E-100.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service