Skip to Content
Merck
All Photos(1)

Key Documents

CF0038

Sigma-Aldrich

Difluoromethyl azide solution

(1.2 M in DME)

Synonym(s):

Azidodifluoromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHF2N3
CAS Number:
MDL number:
UNSPSC Code:
12352125
NACRES:
NA.06
Pricing and availability is not currently available.

form

liquid

Quality Level

concentration

(1.2 M in DME)

SMILES string

[N-]=[N+]=NC(F)F

InChI

1S/CHF2N3/c2-1(3)5-6-4/h1H

InChI key

QPTUUTFPCXREEJ-UHFFFAOYSA-N

Application

Difluoromethyl azide dissolved in DME is a conveniently handled form of this rare fluorinated building block, opening a straightforward access to regioisomerically pure N-difluoromethyl triazoles using the conventional copper catalyzed alkyne-azide cycloaddition or the enamine-mediate azide-ketone [3+2] cycloaddition

Learn more about fluorinated click chemistry

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

26.6 °F

Flash Point(C)

-3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluorinated azides are a new tool that can be used to introduce fluorinated groups to molecules. These molecules can utilize click chemistry to easily add fluorinated groups.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service