Skip to Content
Merck
All Photos(1)

Documents

CDS021585

Sigma-Aldrich

[2-Amino-3-(4-bromobenzoyl)phenyl]acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12BrNO3
CAS Number:
Molecular Weight:
334.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

SMILES string

NC1=C(C=CC=C1C(C2=CC=C(C=C2)Br)=O)CC(O)=O

InChI

1S/C15H12BrNO3/c16-11-6-4-9(5-7-11)15(20)12-3-1-2-10(14(12)17)8-13(18)19/h1-7H,8,17H2,(H,18,19)

InChI key

ZBPLOVFIXSTCRZ-UHFFFAOYSA-N

Gene Information

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Idaira Sánchez-Santos et al.
Experimental eye research, 194, 108020-108020 (2020-03-27)
There is a need to find alternative treatments for MEe. Bromfenac has shown promise in inhibiting the COX-2 enzymatic pathway that partially causes the inflammatory cascade which contributes to the precipitation of ME. However, like other NSAID's, its intraocular half-life
Keith A Walter et al.
Arquivos brasileiros de oftalmologia, 83(1), 55-61 (2019-10-31)
To evaluate the rate of cystoid macular edema development among cataract surgery patients on four different therapeutic regimens. The present study is a retrospective analysis of 5,380 eyes following uncomplicated phacoemulsification at Wake Forest University. The study period went from
Andrés Fernández-Vega Cueto et al.
International ophthalmology, 41(2), 707-717 (2020-10-26)
To assess the effectiveness, safety and predictability of femtosecond laser-assisted cataract surgery (FLACS) in eyes with shallow anterior chamber (AC). This is a prospective consecutive clinical study. All eyes presented an anterior chamber depth (ACD) ≤ 2.1 mm and were submitted to FLACS
Constantine D Georgakopoulos et al.
Retina (Philadelphia, Pa.), 37(2), 388-395 (2016-07-22)
To evaluate the analgesic effect of bromfenac, a topically administered nonsteroidal antiinflammatory agent, in patients undergoing intravitreal injections (IVIs) of anti-vascular endothelial growth factor agents. A single center, prospective, randomized, double-blind, placebo-controlled, cross over interventional study. Patients scheduled to undergo
Chunyue Wang et al.
Medicine, 99(49), e23131-e23131 (2020-12-09)
A cataract is a degenerative change in the optical quality of the lens caused by protein denaturation. Modern medicine is mainly based on surgical treatment. Cataract surgery is often accompanied by severe inflammation, and glucocorticoid therapy has many adverse reactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service