Skip to Content
Merck
All Photos(1)

Key Documents

CDS020611

Sigma-Aldrich

1-Isoindolinone

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$230.00

$230.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
250 MG
$230.00

About This Item

Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

$230.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

SMILES string

O=C1NCC2=C1C=CC=C2

InChI

1S/C8H7NO/c10-8-7-4-2-1-3-6(7)5-9-8/h1-4H,5H2,(H,9,10)

InChI key

PXZQEOJJUGGUIB-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia L Chelius et al.
Fungal genetics and biology : FG & B, 125, 1-12 (2019-01-15)
The protein kinase MpkA plays a prominent role in the cell wall integrity signaling (CWIS) pathway, acting as the terminal MAPK activating expression of genes which encode cell wall biosynthetic enzymes and other repair functions. Numerous studies focus on MpkA
Leixia Chu et al.
Critical reviews in biotechnology, 40(5), 571-589 (2020-04-21)
Compared to terrestrial environments, the oceans harbor a variety of environments, creating higher biodiversity, which gives marine natural products a high occurrence of significant biology and novel chemistry. However, traditional bioassay-guided isolation and purification strategies are severely limiting the discovery
Bruno Perlatti et al.
Molecules (Basel, Switzerland), 25(4) (2020-02-23)
Aspergillus pachycristatus is an industrially important fungus for the production of the antifungal echinocandin B and is closely related to model organism A. nidulans. Its secondary metabolism is largely unknown except for the production of echinocandin B and sterigmatocystin. We
Robin Mesnage et al.
Environmental research, 182, 109116-109116 (2020-02-20)
Glyphosate-based herbicides, such as Roundup®, are the most widely used non-selective, broad-spectrum herbicides. The release of these compounds in large amounts into the environment is susceptible to affect soil quality and health, especially because of the non-target effects on a
Inge Kjærbølling et al.
Fungal genetics and biology : FG & B, 130, 107-121 (2019-06-14)
Filamentous fungi produce a vast number of bioactive secondary metabolites (SMs), some of which have found applications in the pharmaceutical industry including as antibiotics and immunosuppressants. As more and more species are whole genome sequenced the number of predicted clusters

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service