Skip to Content
Merck
All Photos(1)

Key Documents

CDS001518

Sigma-Aldrich

3-Propylphenol

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$77.00

$77.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
250 MG
$77.00

About This Item

Empirical Formula (Hill Notation):
C9H12O
CAS Number:
Molecular Weight:
136.19
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

$77.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

liquid

SMILES string

CCCc1cccc(O)c1

InChI

1S/C9H12O/c1-2-4-8-5-3-6-9(10)7-8/h3,5-7,10H,2,4H2,1H3

InChI key

MPWGZBWDLMDIHO-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Skin Corr. 1C - STOT SE 3

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B Rayaisse et al.
PLoS neglected tropical diseases, 4(3), e632-e632 (2010-03-20)
Field studies were done of the responses of Glossina palpalis palpalis in Côte d'Ivoire, and G. p. gambiensis and G. tachinoides in Burkina Faso, to odours from humans, cattle and pigs. Responses were measured either by baiting (1.) biconical traps
Anibariki Ngonyoka et al.
Journal of vector ecology : journal of the Society for Vector Ecology, 42(1), 34-43 (2017-05-16)
Human activities modify ecosystem structure and function and can also alter the vital rates of vectors and thus the risk of infection with vector-borne diseases. In the Maasai Steppe ecosystem of northern Tanzania, local communities depend on livestock and suitable
Katrin Matheis et al.
Journal of agricultural and food chemistry, 67(5), 1484-1494 (2018-12-19)
The sensomics concept was applied to two steam-treated rapeseed oils, one eliciting a fishy off-flavor (OF) and the other the desired aroma attributes (PC = positive control). After screening the most important odorants via comparative aroma extract dilution analysis, identification
Yuhe Liao et al.
Science (New York, N.Y.), 367(6484), 1385-1390 (2020-02-15)
The profitability and sustainability of future biorefineries are dependent on efficient feedstock use. Therefore, it is essential to valorize lignin when using wood. We have developed an integrated biorefinery that converts 78 weight % (wt %) of birch into xylochemicals.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service