Skip to Content
Merck
All Photos(1)

Key Documents

CDS001457

Sigma-Aldrich

3,5-Ditert-butyl-4-hydroxybenzaldehyde

AldrichCPR

Synonym(s):

2,6-Di-tert-Butyl-4-formylphenol, 3,5-Di-tert-butyl-4-hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$102.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
250 MG
$102.00

About This Item

Empirical Formula (Hill Notation):
C15H22O2
CAS Number:
Molecular Weight:
234.33
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:

$102.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

SMILES string

CC(C)(C)c1cc(C=O)cc(c1O)C(C)(C)C

InChI

1S/C15H22O2/c1-14(2,3)11-7-10(9-16)8-12(13(11)17)15(4,5)6/h7-9,17H,1-6H3

InChI key

DOZRDZLFLOODMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lyndsey F Meyer et al.
Journal of pharmaceutical analysis, 9(3), 163-169 (2019-07-13)
Here we have presented a sensitive and selective LC-MS/MS method for the quantification of tyrphostin A9, which is a selective inhibitor for platelet derived growth factor receptor tyrosine kinase and has been investigated in vitro as a potent oxidative phosphorylation
Bruna Lannuce Silva Cabral et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 107, 1-15 (2017-06-20)
This study shows the design, synthesis and antitumoral potential evaluation of a novel chalcone-like compound, (E)-3- (3, 5-di-ter-butyl-4-hydroxyphenyl)-1- (4-hydroxy-3-methoxyphenyl) prop-2-en-1-one [LQFM064) (4)], against human breast adenocarcinoma MCF7 cells. Some toxicological parameters were also investigated. LQFM064) (4) exhibited cytotoxic activity against
Bo Tao et al.
Journal of analytical methods in chemistry, 2020, 8869576-8869576 (2020-07-14)
The infusion bag is mainly made up of polyolefin polymer. Antioxidants are usually added to these polymer materials in the production process to prevent the materials from aging and enhance the stability of the materials. Because of the potential harm
Paula Vera et al.
Talanta, 188, 750-762 (2018-07-22)
Migration of non volatile compounds from twenty six PP films used as food contact materials has been studied in four simulants (ethanol 95% and 10%, acetic acid 3% and Tenax ®) and analyzed by UPLC-MS/QTOF. Seventy six compounds have been
Alberto Camacho et al.
Lipids in health and disease, 17(1), 128-128 (2018-05-31)
Excessive subcutaneous adiposity in obesity is associated to positive white adipocyte tissue (WAT) differentiation (adipogenesis) and WAT expandability. Here, we hypothesized that supplementation with the insulin inhibitor and mitochondrial uncoupler, Tyrphostin (T-AG17), in vitro and in vivo inhibits adipogenesis and

Questions

  1. Is it a secondary standard/ analytical standard?

    1 answer
    1. This product is not suitable for use as a standard. The item is an AldrichCPR product and part of the rare chemical library. No analytical testing is performed on this product and the structure is not determined. Please read the description section of this product page carefully:
      NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service