Skip to Content
Merck
All Photos(1)

Key Documents

CDS001277

Sigma-Aldrich

Calcium chromate

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$85.40

$85.40

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 G
$85.40

About This Item

Empirical Formula (Hill Notation):
CaCrO4
CAS Number:
Molecular Weight:
156.07
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

$85.40

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

reaction suitability

core: chromium
reagent type: catalyst

storage temp.

−20°C

SMILES string

[Ca++].[O-][Cr]([O-])(=O)=O

InChI

1S/Ca.Cr.4O/q+2;;;;2*-1

InChI key

RFAFBXGYHBOUMV-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Patti C Zeidler-Erdely et al.
Toxicology letters, 334, 60-65 (2020-09-23)
Iron oxides are Group 3 (not classifiable as to its carcinogenicity to humans) according to the International Agency for Research on Cancer (IARC). Occupational exposures during iron and steel founding and hematite underground mining as well as other iron predominant
Haibo Zhang et al.
Bulletin of environmental contamination and toxicology, 103(1), 3-9 (2018-07-20)
The chromate ore process residues (COPR) polluted soil was physically separated into coarse sand (2.000-0.425 mm), fine sand (0.425-0.053 mm) and silt to clay (< 0.053 mm) fractions. The Cr speciation was characterized by synchrotron based micro X-ray fluorescence (µ-XRF) and micro X-ray absorption

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service