Skip to Content
Merck
All Photos(1)

Key Documents

CDS000369

Sigma-Aldrich

N,N-Bis(2-hydroxyethyl)dodecylamine

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$77.00

$77.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
500 MG
$77.00

About This Item

Empirical Formula (Hill Notation):
C16H35NO2
Molecular Weight:
273.45
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

$77.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

SMILES string

CCCCCCCCCCCCN(CCO)CCO

InChI

1S/C16H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-17(13-15-18)14-16-19/h18-19H,2-16H2,1H3

InChI key

NKFNBVMJTSYZDV-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunfeng Chai et al.
Rapid communications in mass spectrometry : RCM, 33(10), 969-977 (2019-03-13)
The incredible sensitivity of the modern mass spectrometry instrument enables scientists to detect a large number of molecules ranging from small organic compounds to biological macromolecules. However, the same sensitivity often throws up challenges with respect to background interferences and
Mahdiyeh Otoukesh et al.
Food chemistry, 321, 126720-126720 (2020-04-11)
Migration of N,N-Bis(2-hydroxyethyl) alkyl(C8-C18)amines from five different polypropylene capsules to Tenax® and coffee powder have been studied. A single step extraction-cleanup procedure using salting out liquid-liquid extraction (SALLE) method followed by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) was applied.
Wellington da Silva Oliveira et al.
Food research international (Ottawa, Ont.), 123, 529-537 (2019-07-10)
Baby bottles made of polypropylene, Tritan® and silicone were evaluated regarding the migration of non-volatile compounds using UPLC-QTOF-MS. Twenty-seven compounds were identified. In all polypropylene samples the migration of 2.2'-(tridecylimino)bis-ethanol and derivatives thereof were detected in concentrations below the specific
Paula Vera et al.
Talanta, 188, 750-762 (2018-07-22)
Migration of non volatile compounds from twenty six PP films used as food contact materials has been studied in four simulants (ethanol 95% and 10%, acetic acid 3% and Tenax ®) and analyzed by UPLC-MS/QTOF. Seventy six compounds have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service