Skip to Content
Merck
All Photos(1)

Key Documents

CDS000104

Sigma-Aldrich

Phlorizin hydrate

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$77.00

$77.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
100 MG
$77.00

About This Item

Empirical Formula (Hill Notation):
C21H26O11
Molecular Weight:
454.42
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

$77.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

crystals

SMILES string

O.OC[C@H]1O[C@@H](Oc2cc(O)cc(O)c2C(=O)CCc3ccc(O)cc3)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H24O10.H2O/c22-9-16-18(27)19(28)20(29)21(31-16)30-15-8-12(24)7-14(26)17(15)13(25)6-3-10-1-4-11(23)5-2-10;/h1-2,4-5,7-8,16,18-24,26-29H,3,6,9H2;1H2/t16-,18-,19+,20-,21-;/m1./s1

InChI key

KOIIMADCRCFGOA-NVADMLJHSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kebede Taye Desta et al.
Biomedical chromatography : BMC, 30(2), 225-231 (2015-06-23)
Agastache rugosa Kuntze (Korean mint) is used as a spice and in folk medicine in East Asia. The present study identified a total of 18 polyphenols from the flower, leaf, stem and roots of this plant using high-performance liquid chromatography-tandem
Hayat Ouassou et al.
Evidence-based complementary and alternative medicine : eCAM, 2018, 9589472-9589472 (2018-09-20)
Many medicinal plants around the world are used for therapeutic purposes against several diseases, including diabetes mellitus. Due to their composition of natural substances that are effective and do not represent side effects for users, unlike synthetic drugs, in this
Ravi Kumar et al.
Life sciences, 244, 117346-117346 (2020-01-25)
Emerging studies have shown that application of low concentration of bioactive phytomolecules can confer anti-proliferative effects on tumour cells by inducing senescence pathways. The alkaloid berberine is recognized for its anti-cancer attributes but its potential to induce senescence in tumour
Aline David-Silva et al.
Diabetes, metabolic syndrome and obesity : targets and therapy, 13, 739-751 (2020-04-02)
NAFLD is a hepatic component of type 2 diabetes mellitus (T2D), in which impaired hepatic glucose production plays an important role. Inhibitors of sodium glucose transporter 2 (SGLT2) reduce glycemia and exert beneficial effects on diabetic complications. Recently, dual SGLT1/2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service