Key Documents
CAD9880
Dissolution cannulae
gauge 14, 316 stainless steel hub, L 4 3/8 in.
Select a Size
1 EA
$1,040.00
$1,040.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,040.00
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
$1,040.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
316 stainless steel hub
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 9880
L
4 3/8 in.
gauge
14
General description
Ideal for drug dissolution testing. Drug dissolution testing is routinely used in the pharmaceutical industry to provide critical release information for quality control purposes, in new drug development, and to predict in vivo drug release profiles. These cannula feature 316 stainless steel hub and tubing for maximum resistance to chemical reaction.
Choose from one of the most recent versions:
Certificates of Analysis (COA)