C97202
1,2-Epoxycyclododecane
95%
Synonym(s):
Cyclododecene oxide (cis+trans)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H22O
CAS Number:
Molecular Weight:
182.30
Beilstein:
106689
EC Number:
MDL number:
UNSPSC Code:
12162002
Assay
95%
refractive index
n20/D 1.48 (lit.)
density
0.939 g/mL at 25 °C (lit.)
SMILES string
C1CCCCCC2OC2CCCC1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service