Skip to Content
Merck
All Photos(1)

Key Documents

C73354

Sigma-Aldrich

3-Methylbenzyl chloride

98%

Synonym(s):

α-Chloro-m-xylene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$100.00
100 G
$270.00

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$100.00
100 G
$270.00

About This Item

Linear Formula:
CH3C6H4CH2Cl
CAS Number:
Molecular Weight:
140.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.535 (lit.)

bp

195-196 °C (lit.)

density

1.064 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1cccc(CCl)c1

InChI

1S/C8H9Cl/c1-7-3-2-4-8(5-7)6-9/h2-5H,6H2,1H3

InChI key

LZBOHNCMCCSTJX-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matthew C Bernier et al.
Journal of mass spectrometry : JMS, 50(7), 891-898 (2015-09-10)
Inorganic metal oxides have shown potential as matrices for assisting in laser desorption ionization with advantages over the aromatic acids typically used. Rhenium and tungsten oxides are attractive options due to their high work functions and relative chemical inertness. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service