Skip to Content
Merck
All Photos(4)

Documents

C44400

Sigma-Aldrich

2-Amino-4-chlorophenol

97%

Synonym(s):

5-Chloro-2-hydroxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(Cl)OH
CAS Number:
Molecular Weight:
143.57
Beilstein:
774859
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050738
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

136-141 °C (lit.)

SMILES string

Nc1cc(Cl)ccc1O

InChI

1S/C6H6ClNO/c7-4-1-2-6(9)5(8)3-4/h1-3,9H,8H2

InChI key

SWFNPENEBHAHEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Marcelo N N Vieira et al.
Cell biochemistry and biophysics, 44(3), 549-553 (2006-05-09)
Protein amyloid aggregation is associated with a number of important human pathologies, but the precise mechanisms underlying the toxicity of amyloid aggregates are still incompletely understood. In this context, drugs capable of blocking or interfering with the aggregation of amyloidogenic
S Ramalingam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 104, 337-351 (2013-01-01)
The spectra (FT-IR and FT-Raman) of the present compound; 2-amino-4-chlorophenol (2A4CP) were recorded in the range of 4000-100 cm(-1). All the computational calculations were made in the ground state using the HF and DFT (B3LYP and B3PW91) methods with 6-31++G(d,p)
Industrial contact dermatitis due to nitro and amino derivatives. 2nd report: experimental study of cross sensitivities of amino derivatives.
S Naniwa
The Journal of dermatology, 9(5), 367-373 (1982-10-01)
International journal of toxicology, 23 Suppl 2, 1-22 (2004-10-30)
Each of these ingredients function as hair colorants. 5-Amino-4-Chloro-o-Cresol and 5-Amino-6-Chloro-o-Cresol are identified as oxidative hair dyes, that is, they are combined with an oxidizing agent before being applied to the hair. 6-Amino-m-Cresol, 6-Amino-o-Cresol, 4-Amino-m-Cresol, and 5-Amino-4-Chloro-o-Cresol are used in
Kazunori Yamazaki et al.
Journal of occupational health, 51(3), 249-260 (2009-04-25)
This study was carried out to clarify the subchronic and chronic toxicity, and carcinogenicity of 2-amino-4-chlorophenol(ACP). Carcinogenicity, and chronic and subchronic toxicity of ACP were examined by feeding 10 rats of both sexes ACP-containing diet at a dose level of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service