Skip to Content
Merck
All Photos(2)

Key Documents

C2004

Sigma-Aldrich

Choline dihydrogencitrate salt

≥98%

Synonym(s):

Salts of (2-Hydroxyethyl)trimethylammonium

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$160.00

$160.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$160.00

About This Item

Empirical Formula (Hill Notation):
C11H21NO8
CAS Number:
Molecular Weight:
295.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$160.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥98%

form

powder

storage temp.

2-8°C

SMILES string

C[N+](C)(C)CCO.OC(=O)CC(O)(CC([O-])=O)C(O)=O

InChI

1S/C6H8O7.C5H14NO/c7-3(8)1-6(13,5(11)12)2-4(9)10;1-6(2,3)4-5-7/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);7H,4-5H2,1-3H3/q;+1/p-1

InChI key

WRPUOFKIGGWQIJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Natural deep eutectic solvents for enzyme stabilization: A study investigated the role of natural deep eutectic solvents, including Choline dihydrogencitrate, in enhancing the thermostability of Humicola insolens cutinase. This approach could be pivotal for improving enzyme performance in various biotechnological applications (Romano et al., 2023).

Biochem/physiol Actions

Acyl group acceptor

Substrates

Choline acetyltransferase substrate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service