Skip to Content
Merck
All Photos(1)

Key Documents

C110507

Sigma-Aldrich

Cyclooctene oxide

99%

Synonym(s):

9-Oxabicyclo[6.1.0]nonane, Epoxycyclooctane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$172.00

$172.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$172.00

About This Item

Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20
Beilstein:
103543
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$172.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

Quality Level

Assay

99%

form

crystals

bp

55 °C/5 mmHg (lit.)

mp

53-56 °C (lit.)

SMILES string

C1CCCC2OC2CC1

InChI

1S/C8H14O/c1-2-4-6-8-7(9-8)5-3-1/h7-8H,1-6H2

InChI key

MELPJGOMEMRMPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Tang et al.
Chemistry, an Asian journal, 14(23), 4375-4382 (2019-10-28)
The efficient chemical conversion of carbon dioxide (CO2 ) into value-added fine chemicals is an intriguing but challenging route in sustainable chemistry. Herein, a hollow-structured bimetallic zeolitic imidazole framework composed of Zn and Co as metal centers (H-ZnCo-ZIF) has been
Yun Wang et al.
Materials (Basel, Switzerland), 12(20) (2019-10-12)
Catalyzed organic solvent-free (ep)oxidation were achieved using H3PM12O40 (M = Mo or W) complexes ionically grafted on APTES-functionalized nano-silica beads obtained from straightforward method (APTES = aminopropyltriethoxysilane). Those catalysts have been extensively analyzed through morphological studies (Dynamic Light Scattering (DLS)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service