Skip to Content
Merck
All Photos(1)

Key Documents

C109606

Sigma-Aldrich

Cyclooctanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H15OH
CAS Number:
Molecular Weight:
128.21
Beilstein:
1848166
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

refractive index

n20/D 1.486 (lit.)

bp

106-108 °C/22 mmHg (lit.)

mp

14-15 °C (lit.)

density

0.974 g/mL at 25 °C (lit.)

SMILES string

OC1CCCCCCC1

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service