Key Documents
BR94915
BRAND® standard ground joint funnel
funnel O.D. 80 mm
Synonym(s):
Funnels, Standard ground joint funnels
Select a Size
5 EA
$60.60
$60.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
5 EA
$60.60
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
$60.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
Joint
joint ST/NS29/32
packaging
pack of 5 ea
manufacturer/tradename
BRAND 94915
funnel O.D.
80 mm
Related Categories
General description
Half-round body. Stem fits standard ground sockets. Ideal for filling liquids or powders into multi-neck flasks.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)