Key Documents
BR90252
BRAND® ETFE beaker with spout, low form
volume 400 mL, with graduation
Synonym(s):
Beakers, Plastics
Select a Size
1 EA
$90.40
$90.40
Estimated to ship onApril 14, 2025
Select a Size
Change View
1 EA
$90.40
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
$90.40
Estimated to ship onApril 14, 2025
material
ETFE
feature
with graduation
manufacturer/tradename
BRAND 90252
volume
400 mL
General description
Made from high-quality ETFE (ethylene-tetrafluoroethylene), this translucent beaker offers excellent chemical resistance, making it ideal for handling various substances in laboratory settings. The low form provides stability and ease of use, allowing for efficient mixing and pouring of liquids. Clearly marked graduations facilitate accurate measurement of liquids, ensuring precision in experiments and formulations. The beaded rim enhances strength and provides a comfortable grip, making handling safer and more efficient.
Other Notes
Browse all plastic beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)