Merck
All Photos(4)
Key Documents
BR81167
BRAND® burette lateral stopcock
for burette capacity 25+50 mL, with spare key, PTFE key
Synonym(s):
Accessories, Burette stopcock, Burettes, Stopcocks
Select a Size
1 EA
$58.00
$58.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$58.00
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
$58.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
PTFE key
amber
packaging
pack of 1 ea
manufacturer/tradename
BRAND 81167
Related Categories
General description
BRAND burettes are always calibrated to deliver (TD, Ex).
Waiting times:
BLAUBRAND® - waiting time 30 sec. SILBERBRAND - no waiting time
A waiting time is usually not necessary if a titration is performed (see DIN EN ISO 385).
All BLAUBRAND® burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)