Skip to Content
Merck

Skip To

BR781663

BRAND® microplate BRANDplates®, pureGrade

size 96 wells, size 350 μL, conical bottom, transparent

Synonym(s):

BRANDplates®, Microplates, pureGradeS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 ea
Check Cart for Availability
$261.00

About This Item

NACRES:
NB.15
UNSPSC Code:
41121800
Material:
conical bottom, colorless wells, polystyrene
Size:
350 μL , 96 wells

$261.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

conical bottom, colorless wells, polystyrene

sterility

sterile

feature

lid, skirt (C-bottom)

manufacturer/tradename

BRAND 781663

packaging

pack of 50 ea, pkg of (individually packaged)

size

350 μL , 96 wells

working volume

350 μL

color

transparent

suitability

suitable for (bacteriological assays)

binding type

non-treated surface

General description

The BRAND® Microplate BRANDplates® PureGrade features a non-treated surface, making it particularly well-suited for bacteriological assays. Sterile according to ISO 11137 standards, this microplate ensures a contamination-free environment for sensitive experiments. It is classified as BIO-CERT® CELL CULTURE QUALITY, guaranteeing that it is free from human DNA, RNase, DNase, and pyrogens, and is non-cytotoxic. This reliable microplate is an essential tool for researchers conducting microbiological studies, providing consistent performance and safety in cell culture applications.

Features and Benefits

  • Optimal surfaces for optimal results
  • Reliable quality from the cleanroom
  • Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BR781668BR781671BR781660
suitability

suitable for (bacteriological assays)

suitability

suitable for (bacteriological assays)

suitability

suitable for (bacteriological assays)

suitability

suitable for (bacteriological assays)

size

350 μL , 96 wells

size

350 μL , 96 wells

size

330 μL , 96 wells

size

330 μL , 96 wells

material

colorless wells, polystyrene , conical bottom

material

black wells, flat bottom, polystyrene

material

black wells, flat bottom, polystyrene

material

colorless wells, polystyrene , round bottom

manufacturer/tradename

BRAND 781663

manufacturer/tradename

BRAND 781668

manufacturer/tradename

BRAND 781671

manufacturer/tradename

BRAND 781660

color

transparent

color

black

color

black, with transparent bottom

color

transparent

working volume

350 μL

working volume

350 μL

working volume

330 μL

working volume

330 μL


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
BR781663-50EA04061834131892