BR73724
BRAND® filter flask, glass, with lateral socket
capacity 1000 mL
Synonym(s):
Bottles, Filter flask, Glass
Select a Size
$119.00
Select a Size
About This Item
$119.00
Recommended Products
material
borosilicate glass
flat bottom flask
Joint
joint ST/NS neck33.7 mm
packaging
pkg of 1 ea
manufacturer/tradename
BRAND 73724
bottle capacity
1000 mL
capacity
1000 mL
funnel capacity
1000 mL
neck diam.
45 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service