BR705384
Individual stand for pipette Transferpette® electronic + -8-12 electronic
Select a Size
$306.00
Estimated to ship onMay 28, 2025
Select a Size
About This Item
$306.00
Estimated to ship onMay 28, 2025
Recommended Products
material
PBT
PC
packaging
pack of 1 ea
manufacturer/tradename
BRAND 705384
General description
Wall/rack mounts are available for mechanical single- and multichannel Transferpette® S pipettes. The single stand for the Transferpette® S mechanical pipette is usually used for the multichannel pipette. Single stands are also available for the Transferpette® electronic pipette - one stand for pipettes up to 1000 µl and another for the 5 mL pipette. The benchtop rack (carousel) stand holds up to six Transferpette® S or Transferpette® S -8/-12 pipettes, saving considerable space. Three Transferpette® electronic pipettes (single channel, up to 1000 μl) can be charged simultaneously in the charging stand. Individual power supplies no longer needed!
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)