Key Documents
BR701370
BRAND® sealing film for microplates
self-adhesive, suitable for (for automation), polypropylene
Synonym(s):
Accessories, PCR, Plates, Sealing films
Select a Size
50 EA
$259.00
$259.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
50 EA
$259.00
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
$259.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
film (PP/PE-HD)
polypropylene
sterility
non-sterile
feature
piercable
self-adhesive
packaging
pack of 50 ea
manufacturer/tradename
BRAND 701370
suitability
suitable for (for automation)
General description
Due to the increasing number of analysis methods, the number of samples processed is growing steadily. This means that high-throughput analyses are gaining significance. In these automated processes, the films must not only protect the samples, but also comply with the mechanical requirements of automation.
Features and Benefits
  • Pierceability
  • Adhesive-free zones for contamination-free tasks
  • Tight seal for minimizing evaporation
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)