Key Documents
BR701358
BRAND® mat cover for deep well plate
volume 0.5 mL
Synonym(s):
Accessories, Cover mats, Deep-well Plates, Sample Storage
Select a Size
50 EA
$597.00
$597.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
50 EA
$597.00
About This Item
UNSPSC Code:
41122102
NACRES:
NB.22
$597.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
sterility
non-sterile
manufacturer/tradename
BRAND 701358
volume
0.5 mL
General description
These cover mats reduce the maximum volume of wells but minimize evaporation.They can be used in conjunction with adhesive sealing films for added protection and containment.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)