Key Documents
BR476900
BRAND® slide box
Holds 5 or 10 x slides 76x26 mm
Synonym(s):
Microscope slides, Microscopy, Slide box, Staining, Staining Accessories
Select a Size
10 EA
$126.00
$126.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
10 EA
$126.00
About This Item
UNSPSC Code:
41123302
NACRES:
MA.01
$126.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
feature
Holds 5 or 10 x slides 76x26 mm
packaging
pack of 10 ea
manufacturer/tradename
BRAND 476900
General description
Sturdy design. Screw lid with dealing wire lug. Ideal for storing or shipping 5 thick or 10 thin slides. Slides protrude 10 mm above the box, to facilitate removal.
For slides, size (in, mm) 76 x 26. Numbered slots in base. Lid with index card.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Jiasheng Wang et al.
Bioscience, biotechnology, and biochemistry, 84(11), 2215-2227 (2020-07-24)
This study explored the mechanism of NEAT1 in sepsis-induced AKI rats. Cecal ligation punctures (CLP)-induced AKI rats were injected with siRNA-NEAT1 lentivirus. Kidney histopathology and apoptosis were evaluated via hematoxylin-eosin and TUNEL staining, respectively. ELISA determined the levels of Blood