Key Documents
BR36975
BRAND® volumetric flask, USP, BLAUBRAND®
capacity 20 mL, joint: ST/NS 10/19, glass stopper
Synonym(s):
BRAND® volumetric flask BLAUBRAND® USP conformity certified, BLAUBRAND®, USP, Volumetric flasks
Select a Size
2 EA
$79.80
$79.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
2 EA
$79.80
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$79.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
glass flask
glass stopper
Joint
joint: ST/NS10/19
feature
accuracy class A
packaging
pack of 2 ea
manufacturer/tradename
BRAND 36975
capacity
20 mL
General description
The BRAND® volumetric flask, USP, BLAUBRAND® is constructed from high-quality Boro 3.3 glass and adheres to DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring precise liquid measurement for various laboratory applications. Each flask is accompanied by a USP batch certificate, providing assurance of compliance with stringent quality standards. This volumetric flask is an essential tool for accurate analytical work, making it suitable for laboratories that require reliable and certified liquid handling solutions.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)