BR36954
BRAND® volumetric flask BLAUBRAND® USP conformity certified
capacity 2000 mL, neck joint: ST/NS 29/32, polypropylene stopper
Synonym(s):
BLAUBRAND®, USP, Volumetric flasks
Select a Size
$140.00
Select a Size
About This Item
$140.00
Recommended Products
material
borosilicate glass
polypropylene stopper
Joint
joint: ST/NS neck29/32
feature
accuracy class A
packaging
pack of 2
manufacturer/tradename
BRAND 36954
capacity
2000 mL
Related Categories
General description
Features and Benefits
- BLAUBRAND® 2000 mL volumetric flasks offer precise measurement of liquids with high accuracy even for partial volumes.
- Computer-controlled systems guarantee the possible precision for fully automated production processes.
- Individually calibrated for the best technical perfection.
- USP volumetric instruments comply with the Class A error limits required by the United States Pharmacopeia.
- The flask can be heated up to 250°C in a drying cabinet or sterilizer without any permanent volume changes.
- The use of specially developed high-quality printing inks ensures a long service life.
- The blue enamel used for BLAUBRAND® volumetric instruments offers an excellent color contrast. It represents the optimum combination of resistance and readability.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)