Skip to Content
Merck
All Photos(1)

Key Documents

BR30611

BRAND® USP BLAUBRAND® volumetric pipettes, one-mark

capacity 9 mL, class AS, USP, DE-M

Synonym(s):

BLAUBRAND®, Bulb pipettes, USP

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
$156.00

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
6 EA
$156.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

AR-Glass

feature

class AS USP, DE-M
DE-M marking

packaging

pack of 6 ea

manufacturer/tradename

BRAND 30611

capacity

9 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The BRAND® USP BLAUBRAND® volumetric pipettes, one-mark, are crafted from high-quality AR-Glas® and comply with DIN EN ISO 648 standards. These pipettes are adjusted to "Ex" and feature a wait time of 5 seconds for precise liquid delivery. Each pipette includes a USP batch certificate, ensuring quality assurance and traceability. This reliable volumetric pipette is essential for accurate liquid measurement in various laboratory applications, making it a valuable tool for precise analytical work.

Features and Benefits

  • BRAND® USP BLAUBRAND® volumetric bulb pipette, 1-mark is used for the accurate measurement of instruments for precision analyses.
  • Marks and inscriptions in high contrast blue enamel.        
  • Manufactured according to the DIN EN ISO 648 standards.        
  • Adjusted to "Ex". Wait time 5 seconds.        
  • Includes USP batch certificate.        
  • Also available with USP individual certificate upon request.

Legal Information

AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page contains detailed information on working with and handling pipettes and pipetting aids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service