Key Documents
BR122620
BRAND® screw cap
thread for GL 25, integral glass ring
Synonym(s):
Accessories, Bottles, Screw caps
Select a Size
10 EA
$49.30
$49.30
Estimated to ship onApril 17, 2025
Select a Size
Change View
10 EA
$49.30
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$49.30
Estimated to ship onApril 17, 2025
material
polypropylene
description
for laboratory bottles GL 25
packaging
pack of 10 ea
manufacturer/tradename
BRAND 122620
fitting
thread for GL 25
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)