Key Documents
BASMF2064
Sigma-Aldrich
Replacement frit for Reference Electrodes
Synonym(s):
Reference Electrode Frit
Select a Size
1 EACH
$325.00
$325.00
PRICE WITHOUT NATIONAL TAXES
Available to ship onApril 28, 2025Details
Request a Bulk Order
Select a Size
Change View
1 EACH
$325.00
About This Item
UNSPSC Code:
12352300
NACRES:
NA.23
$325.00
PRICE WITHOUT NATIONAL TAXES
Available to ship onApril 28, 2025Details
Request a Bulk Order
packaging
pack of 5 ea
General description
1/8" chunks porous CoralPor with 1/2" tubes PTFE heat shrink
Legal Information
Product of Bioanalytical Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. this kit includes 2 pieces in total?
    1 answer
    1. This product contains a total of 5 pieces.
      Helpful?
  2. Are these frits suitable for Hg/HgO (1M KOH) electrodes?
    1 answer
    1. No, the MF-2064 CoralPor tips will dissolve in an alkaline solution.
      Helpful?