Skip to Content
Merck
All Photos(2)

Key Documents

BASMF2060

Sigma-Aldrich

Electrode Polishing Kit, PK-4

Synonym(s):

PK-4 Electrode Polish

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$463.00

$463.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
$463.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

$463.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

General description

Kit includes:
  • Electrode modifier- fine grit polishing pads (5)
  • Diamond polishing pads (white, nylon) (10)
  • Alumina polishing pads (tan, velvet) (5)
  • Polishing glass plate (2)
  • Electrode modifier - coarse diamond polish - 15 um (1)
  • Electrode modifier - fine diamond polish - 3 um (1)
  • Electrode modifier - ultrafine diamond polish - 1 um (1)
  • Electrode modifier - polishing alumina (1)

Application

The electron transfer kinetics, and hence the current response, of redox-active species can be dramatically affected by the condition of the surface of the working electrode. Pretreatment of the electrode surface may therefore be necessary in order to obtain reproducible results.

If rinsing the electrode surface with water and/or methanol is insufficient to remove adsorbed species and restore the working electrode to prime condition, then hand polishing may be required using the PK-4 polishing kit. The polish suspensions are applied to fabric pads attached to glass plates. The progression of polishing should always proceed from coarse to fine materials, with thorough rinsing and a change of pads between each step.

Legal Information

Product of Bioanalytical Systems, Inc.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service