Key Documents
BAH379400000
SP Bel-Art® FEP Coated Steel Forceps
curved, sharp point PTFE-coated stainless steel forceps, L 3.875 in. (98 mm) , tip coating
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
Pricing and availability is not currently available.
material
curved, sharp point PTFE-coated stainless steel forceps
packaging
pkg of 2 ea (48/case)
manufacturer/tradename
Bel-Art H37940-0000
L
3.875 in. (98 mm) , tip coating
General description
The exceptionally fine tips of these forceps are coated with brown Fluo-Kem® PTFE FEP.
Features and Benefits
  • Complies with FDA regulations covering direct contact with food.
  • Serrated, curved tips
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Fluo-kem is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Can these SP Bel-Art® FEP Coated Steel Forceps be autoclaved?
    1 answer
    1. This is a product of SP Bel-Art. The manufacturer states that this item is autoclavable.
      Helpful?