Key Documents
BAH367190000
SP Bel-Art® Quaverette Micro-Blade Vibrating Spatula
L 8.25 in. (21 cm), brown FEP blade
Synonym(s):
spoon
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
Pricing and availability is not currently available.
material
brown FEP blade
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H36719-0000
L
8.25 in. (21 cm)
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
BAH367180000BAH367140000BAH367150000
material
brown FEP blade
material
stainless steel blade
material
stainless steel blade
material
brown FEP blade
manufacturer/tradename
Bel-Art H36719-0000
manufacturer/tradename
Bel-Art H36718-0000
manufacturer/tradename
Bel-Art H36714-0000
manufacturer/tradename
Bel-Art H36715-0000
L
8.25 in. (21 cm)
L
-
L
-
L
7 in. (17.78 cm)
General description
Make accurate additions of precious materials as needed. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving you control over transfer from spatula to weighing dish.
Features and Benefits
Available with a plain stainless steel blade or with a brown FEP coating
Coating complies with FDA regulations governing direct food contact applications; reduces transfer losses
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)