Key Documents
BAH188620000
Scienceware® tiered test tube rack
Holds 16 x 10-14 mm tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
material
high-density polypropylene
feature
Holds 16 x 10-14 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H18862-0000
L × W × H
23.9 cm × 6.4 cm × 6.6 cm
size
330x330x44 mm
General description
Compact, test tube rack takes up very little space and keeps up to 16 tubes upright and organized. Tiered for easier viewing.
  • 16 place rack holds tubes 10-14mm
  • Double tiered rack with solid bottom platform, keeps tubes stable
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)