Documents
BAF249870000
Beta-Safe storage box for β-emitters
W × L × H 25.4 cm (10 in.) × 15.24 cm (6 in.) × 17.8 cm (7 in.)
About This Item
UNSPSC Code:
41123300
NACRES:
NB.35
material
acrylic
feature
Radioactive materials such as P-32, S-35 and C-14
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F24987-0000
W × L × H
25.4 cm (10 in.) × 15.24 cm (6 in.) × 17.8 cm (7 in.)
General description
Rubber feet safeguards bench and reduce movement. Lock and cable or chain not included. Thick 9.5 mm (3/8 in.) translucent, acrylic safeguards against radioactive isotopes.
  • Use to hold 32-P, 35-S and 14-C
  • Hinged lid has built in lock hasp to shelter contents
  • Side mounted lock tabs allow box to be fixed to objects with a cable
Legal Information
Beta-Safe is a trademark of SP Scienceware, a division of SP