Skip to Content
Merck
All Photos(1)

Documents

BAF116460627

Scienceware® safety labeled wash bottles

label, Sodium Hypochlorite (Bleach), wide-mouth yellow cap, capacity 500 mL (16 oz)

Synonym(s):

dispensing bottle, safety bottles, squeeze bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

material

low-density polyethylene
wide-mouth yellow cap

feature

label Sodium Hypochlorite (Bleach)

packaging

pack of 6 ea

manufacturer/tradename

Bel-Art F11646-0627

capacity

500 mL
500 mL (16 oz)

color

opaque

Looking for similar products? Visit Product Comparison Guide

General description

Bel-Art’s Safety Labeled wash bottles include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, making your workplace a safer place.

Features and Benefits

  • Easy-to-read printed bottles
  • Wide mouth 53mm (2") bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Note: Use LYOB only with solvents compatible with LDPE.

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service