Skip to Content
Merck
All Photos(1)

Key Documents

B89000

Sigma-Aldrich

sec-Butylamine

99%

Synonym(s):

(±)-sec-Butylamine, (±)-2-Aminobutane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$62.60
500 ML
$79.60

$62.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$62.60
500 ML
$79.60

About This Item

Linear Formula:
CH3CH2CH(NH2)CH3
CAS Number:
Molecular Weight:
73.14
Beilstein:
1361345
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$62.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.3928 (lit.)

bp

63 °C (lit.)

mp

−72 °C (lit.)

density

0.724 g/mL at 25 °C (lit.)

SMILES string

CCC(C)N

InChI

1S/C4H11N/c1-3-4(2)5/h4H,3,5H2,1-2H3

InChI key

BHRZNVHARXXAHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service