Skip to Content
Merck
All Photos(1)

Key Documents

B87806

Sigma-Aldrich

4-Butoxyphenol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$95.60

$95.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$95.60

About This Item

Linear Formula:
CH3(CH2)3OC6H4OH
CAS Number:
Molecular Weight:
166.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$95.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

impurities

2% hydroquinone

mp

65-66 °C (lit.)

SMILES string

CCCCOc1ccc(O)cc1

InChI

1S/C10H14O2/c1-2-3-8-12-10-6-4-9(11)5-7-10/h4-7,11H,2-3,8H2,1H3

InChI key

MBGGFXOXUIDRJD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frédéric L P Gabriel et al.
Applied and environmental microbiology, 73(10), 3320-3326 (2007-03-21)
Recently we showed that degradation of several nonylphenol isomers with alpha-quaternary carbon atoms is initiated by ipso-hydroxylation in Sphingobium xenophagum Bayram (F. L. P. Gabriel, A. Heidlberger, D. Rentsch, W. Giger, K. Guenther, and H.-P. E. Kohler, J. Biol. Chem.
A Davydov et al.
Biochemical and biophysical research communications, 258(2), 322-325 (1999-05-18)
The regeneration of the tyrosyl radical in chemically reduced native or p-butoxyphenol-treated radical free forms of mouse ribonucleotide reductase R2 protein has been studied. Chemical reduction has been achieved by treatment with light-activated flavin compounds: deazaflavin, flavin mononucleotide, or deazaflavin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service