Skip to Content
Merck
All Photos(1)

Key Documents

B84807

Sigma-Aldrich

1,4-Butanediol

99%

Synonym(s):

1,4-Butylene glycol, Tetramethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)4OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1633445
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

3.1 (vs air)

Assay

99%

form

liquid

autoignition temp.

698 °F

refractive index

n20/D 1.445 (lit.)

bp

230 °C (lit.)

mp

16 °C (lit.)

density

1.017 g/mL at 25 °C (lit.)

SMILES string

OCCCCO

InChI

1S/C4H10O2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

WERYXYBDKMZEQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ari Karchin et al.
Acta biomaterialia, 7(9), 3277-3284 (2011-06-07)
Electrospinning from a melt, in contrast to from a solution, is an attractive tissue engineering scaffold manufacturing process as it allows for the formation of small diameter fibers while eliminating potentially cytotoxic solvents. Despite this, there is a dearth of
Harry Yim et al.
Nature chemical biology, 7(7), 445-452 (2011-05-24)
1,4-Butanediol (BDO) is an important commodity chemical used to manufacture over 2.5 million tons annually of valuable polymers, and it is currently produced exclusively through feedstocks derived from oil and natural gas. Herein we report what are to our knowledge
Sandra Rinne Dahl et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 885-886, 37-42 (2012-01-10)
The demand of high throughput methods for the determination of gamma-hydroxybutyrate (GHB) and its precursors gamma-butyrolactone (GBL) and 1,4-butane-diol (1,4BD) as well as for pregabalin is increasing. Here we present two analytical methods using ultra-high pressure liquid chromatography (UPLC) and
Anders Hamberg et al.
Chemical communications (Cambridge, England), 48(80), 10013-10015 (2012-09-05)
The enzyme Candida antarctica lipase B was subjected to site directed mutagenesis suggested by molecular modelling. The selectivity for the enzyme increased towards a range of diols over their corresponding monoesters as an effect of the mutations.
Lambert K Sørensen et al.
Forensic science international, 222(1-3), 352-359 (2012-08-25)
A liquid-chromatography-tandem-mass-spectrometry method using pneumatically assisted electrospray ionisation (LC-ESI-MS/MS) was developed for the simultaneous determination of γ-hydroxybutyric acid (GHB), γ-butyrolactone (GBL) and 1,4-butanediol (1,4-BD) in human ante-mortem and post-mortem whole blood. The blood proteins were precipitated using a mixture of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service