Skip to Content
Merck
All Photos(1)

Key Documents

B153

Sigma-Aldrich

BIT

Synonym(s):

1-[2-(N,N-Diethylamino)ethyl]-2-(4-ethoxybenzyl)benzimidazole-5-isothiocyanate, Etonitazenyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28N4OS
CAS Number:
Molecular Weight:
408.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

mp

64-66 °C

storage temp.

−20°C

SMILES string

CCOc1ccc(Cc2nc3cc(ccc3n2CCN(CC)CC)N=C=S)cc1

InChI

1S/C23H28N4OS/c1-4-26(5-2)13-14-27-22-12-9-19(24-17-29)16-21(22)25-23(27)15-18-7-10-20(11-8-18)28-6-3/h7-12,16H,4-6,13-15H2,1-3H3

InChI key

QIIKEXXYYPDZHL-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyler Janovitz et al.
Virology, 511, 40-48 (2017-08-15)
The pathogenic autonomous human parvovirus B19 (B19V) productively infects erythroid progenitor cells (EPCs). Functional similarities between B19V nonstructural protein (NS1), a DNA binding endonuclease, and the Rep proteins of Adeno-Associated Virus (AAV) led us to hypothesize that NS1 may facilitate
Chemogenomic Landscape of
Laura Simon et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(22), 6969-6981 (2017-09-01)
Lisheng Wang et al.
The Journal of experimental medicine, 201(10), 1603-1614 (2005-05-11)
Despite the need for alternative sources of human hematopoietic stem cells (HSCs), the functional capacity of hematopoietic cells generated from human embryonic stem cells (hESCs) has yet to be evaluated and compared with adult sources. Here, we report that somatic
Ulrich Heimhofer et al.
Nature communications, 9(1), 3832-3832 (2018-09-22)
The Cenomanian-Turonian Oceanic Anoxic Event (OAE2; ~94.5 million years ago) represents an episode of global-scale marine anoxia and biotic turnover, which corresponds to one of the warmest time intervals in the Phanerozoic. Despite its global significance, information on continental ecosystem
David J H F Knapp et al.
Nature cell biology, 20(6), 710-720 (2018-05-29)
Elucidation of the identity and diversity of mechanisms that sustain long-term human blood cell production remains an important challenge. Previous studies indicate that, in adult mice, this property is vested in cells identified uniquely by their ability to clonally regenerate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service