Skip to Content
Merck
All Photos(1)

Key Documents

ALD00570

Sigma-Aldrich

1-((4-(Trifluoromethyl)phenyl)sulfonyl)bicyclo[2.1.0]pentane

racemic version, 95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$206.00

$206.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$206.00

About This Item

Empirical Formula (Hill Notation):
C12H11F3O2S
Molecular Weight:
276.27
UNSPSC Code:
12352200
NACRES:
NA.22

$206.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

Application

The following "spring-loaded" electrophile is a convenient reagent for the installation of cyclopentane to primary and secondary amines. This product is one of several synthetically useful reagents reported by Baran and coworkers for the “any-stage” installation of small, strained ring systems.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service