Skip to Content
Merck
All Photos(1)

Key Documents

ALD00350

Sigma-Aldrich

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-methoxy-L-phenylalanine

95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$356.00

$356.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$356.00

About This Item

Empirical Formula (Hill Notation):
C25H23NO5
Molecular Weight:
417.45
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$356.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95% (HPLC)

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

171-176 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OC([C@@H](NC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O)CC4=CC=CC=C4OC)=O

InChI

1S/C25H23NO5/c1-30-23-13-7-2-8-16(23)14-22(24(27)28)26-25(29)31-15-21-19-11-5-3-9-17(19)18-10-4-6-12-20(18)21/h2-13,21-22H,14-15H2,1H3,(H,26,29)(H,27,28)/t22-/m0/s1

InChI key

CBCSNZJHURMDMO-QFIPXVFZSA-N

Application

Unnatrual amino acid was derived from a C-H Activation methodology developed by Jin-Quan Yu and coworkers. The Pd-mediated C-C bond formation can be made in tandem with 2-Methylpyridine (Aldrich 109835).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service