All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H3Cl2FO2S
CAS Number:
Molecular Weight:
229.06
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
95%
form
solid
reaction suitability
reaction type: click chemistry
mp
65-70 °C
SMILES string
O=S(C1=C(Cl)C=CC=C1Cl)(F)=O
InChI
1S/C6H3Cl2FO2S/c7-4-2-1-3-5(8)6(4)12(9,10)11/h1-3H
InChI key
YZHIFNHRRUIOAY-UHFFFAOYSA-N
Application
Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service